Anti-HNRNPD Rabbit Polyclonal Antibody

Supplier: Proteintech
12770-1-AP-150UL
10088-378EA 402 USD
10088-378
Anti-HNRNPD Rabbit Polyclonal Antibody
Antibodies
The heterogeneous nuclear ribonucleoprotein (hnRNP) complexes which provide the substrate for the processing events that pre-mRNAs undergo before becoming functional, translatable mRNAs in the cytoplasm. HNRNPD, also known as AUF1, belongs to the family of AU-binding proteins (AU-BPs) that regulate the cellular half-lives of many mRNAs by directly interacting with an AU-rich element (ARE) located in their 3′ untranslated region .

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human cervical cancer, 1:20-1:200; IF: HepG2, 1:20-1:200

Type: Primary
Antigen: HNRNPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR