Anti-PDE12 Rabbit Polyclonal Antibody

Supplier: Proteintech
12753-1-AP-150UL
10091-992EA 402 USD
10091-992
Anti-PDE12 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: PDE12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR