Anti-EXOC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12751-1-AP-150UL
10094-258EA 459.7 USD
10094-258
Anti-EXOC2 Rabbit Polyclonal Antibody
Antibodies
EXOC2 (exocyst complex component 2), also known as SEC5 and SEC5L1, is a component of the exocyst complex, and is required to mediate RalB-dependent survival signals in transformed cells. The exocyst complex, composed of eight evolutionarily conserved subunits (SEC3, SEC5, SEC6, SEC8, SEC10, SEC15, EXO70, and EXO84), is involved in tethering post-Golgi secretory vesicles to specific plasma membrane domains. The gene of EXOC2 maps to chromosome 6p25.3, and encodes a 924-amino acid protein with an experimentally determined molecular mass of 95-100 kDa. EXOC2 mRNA is widely expressed with highest levels in brain and placenta.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: EXOC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR