Anti-MAP2K6 Rabbit Polyclonal Antibody

Supplier: Proteintech
12745-1-AP-150UL
10090-132EA 453 USD
10090-132
Anti-MAP2K6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Kidney Tissue, 1:500-1:5000; IHC: Human breast cancer Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: MAP2K6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR