Anti-EIF3C Rabbit Polyclonal Antibody

Supplier: Proteintech
12733-1-AP-150UL
10086-368EA 402 USD
10086-368
Anti-EIF3C Rabbit Polyclonal Antibody
Antibodies
Western Blot:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: EIF3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR