Anti-PMP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12717-1-AP-150UL
10092-296EA 402 USD
10092-296
Anti-PMP2 Rabbit Polyclonal Antibody
Antibodies
PMP2 (peripheral myelin protein-2, also known as P2) is one of the major proteins of peripheral myelin and appears to be related to the transport of fatty acids or the metabolism of myelin lipids.P2 constitutes up to 15% of total protein of peripheral nervous system (PNS) myelin. It is also present in small amounts in central nervous system (CNS) myelin, being abundant in spinal cord and brain stem myelin. As a structural protein, P2 is thought to stabilize the myelin membranes.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human gliomas, 1:20-1:200

Type: Primary
Antigen: PMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR