Anti-GCNF Rabbit Polyclonal Antibody

Supplier: Proteintech
12712-1-AP-150UL
10087-446EA 402 USD
10087-446
Anti-GCNF Rabbit Polyclonal Antibody
Antibodies
NR6A1 (Nuclear Receptor Subfamily 6 Group A member), also known as GCNF (Germ Cell Nuclear Factor), is an orphan member of the nuclear receptor gene superfamily. It has been shown to be expressed in the nervous system during development and during specific stages in maturing germ cells of the ovary and testis in the adult, and has probable roles in gametogenesis, neurogenesis, and normal embryonic development during gastrulation. Inactivation of GCNF in mouse results in abnormal posterior development, impaired midbrain development, insufficient closure of the neural tube, and eventual embryonic death. GCNF has been shown to be a repressor of OCT-4 and of the protamine genes and plays a critical role in the control of gene expression during embryogenesis and spermatogenesis. This antibody specifically reacts with the 55kd NR6A1 protein.

Western Blot:PC-3 Cells, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: GCNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR