Anti-NEK11 Rabbit Polyclonal Antibody

Supplier: Proteintech
12710-1-AP-150UL
10091-018EA 459.7 USD
10091-018
Anti-NEK11 Rabbit Polyclonal Antibody
Antibodies
NEK11(Serine/threonine-protein kinase) belongs to the NIMA family of kinases, which are involved in DNA replication and genotoxic stress responses.It controls degradation of CDC25A by directly phosphorylating it on residues whose phosphorylation is required for BTRC-mediated polyubiquitination and degradation.It has 4 isoforms produced by alternative splicing.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: NEK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR