![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | RIN3 |
Antigen Name | Ras and Rab Interactor 3 |
Reactivity | Human, Rat, Mouse |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage Buffer | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
Shipping Temperature | Store at -20°C |