Anti-GABRA3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12708-1-AP-150UL
10086-430EA 459.7 USD
10086-430
Anti-GABRA3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: GABRA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR