Anti-HEBP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12706-1-AP-150UL
10088-144EA 459.7 USD
10088-144
Anti-HEBP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IP: HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: HEBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR