Anti-SGK3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12699-1-AP-150UL
10081-504EA 402 USD
10081-504
Anti-SGK3 Rabbit Polyclonal Antibody
Antibodies
Serum- and glucocorticoid-inducible kinase 3 (SGK3) is a protein kinase of the AGC family of protein kinase A, protein kinase G, and protein kinase C and functions downstream of phosphatidylinositol 3-kinase (PI3K). It is also named as DKFZp781N0293, CISK, SGK2, SGKL and may be involved in the regulation of processes such as cell survival, neuronal excitability, and renal sodium excretion. SGK3 mediates cell IL-3-dependent survival signals . SGK3 has 2 isoforms produced by alternative splicing with the molecular weight of 57 kDa and 53 kDa.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IF:A375 Cells, 1:10-1:100

Type: Primary
Antigen: SGK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR