Anti-Kallikrein 11 Rabbit Polyclonal Antibody

Supplier: Proteintech
12698-1-AP-150UL
10089-092EA 459.7 USD
10089-092
Anti-Kallikrein 11 Rabbit Polyclonal Antibody
Antibodies
KLK11(Kallikrein-11) is also named as PRSS20, TLSP and belongs to the peptidase S1 family and Kallikrein subfamily. It is a multifunctinal protease cleaving several substrates for kallikrein and trypsin and involved in normal physiology processes in bronchus. This full length protein has a signal peptide, a propeptide and four glycosylation sites. It has 4 isoforms produced by alternative splicing with the molecular weight of 31 kDa, 27 kDa,34 kDa and 30 kDa.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF:PC-3 Cells, 1:10-1:100

Type: Primary
Antigen: Kallikrein 11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR