Anti-ZFYVE27 Rabbit Polyclonal Antibody

Supplier: Proteintech
12680-1-AP-150UL
10097-276EA 402 USD
10097-276
Anti-ZFYVE27 Rabbit Polyclonal Antibody
Antibodies
ZFYVE27, also named as Protrudin, is a 411 amino acid protein, which contains 1 FYVE-type zinc finger. ZFYVE27 localizes in the recycling endosome membrane and functions as an upstream inhibitor of RAB11, regulating directional protein transport to the forming neurites. ZFYVE27 is a key regulator of vesicular transport during neurite extension and is involved in neuronal intracellular trafficking in the corticospinal tract.

  • Western Blot: Mouse Thymus Tissue, 1:200-1:2000

Type: Primary
Antigen: ZFYVE27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR