Anti-PPP2R5A Rabbit Polyclonal Antibody

Supplier: Proteintech
12675-2-AP-150UL
10092-720EA 459.7 USD
10092-720
Anti-PPP2R5A Rabbit Polyclonal Antibody
Antibodies
PPP2R5A(protein phosphatase 2, regulatory subunit B (B56), alpha isoform) is also named as PR61alpha and belongs to the phosphatase 2A regulatory subunit B56 family. B56 alpha subunit gene encodes a cytoplasmic phosphoprotein. The B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment. This protein has 2 isoforms produced by alternative splicing with the molecular weight of 54 kDa and 50 kDa.

Western Blot: Human ileum Tissue, 1:1000-1:10000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PPP2R5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR