Anti-BRSK1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12673-1-AP-150UL
10083-694EA 459.7 USD
10083-694
Anti-BRSK1 Rabbit Polyclonal Antibody
Antibodies
BRSK1(Brain-specific serine/threonine-protein kinase 1) is also named as KIAA1811, SAD1, SADB and belongs to the SNF1 subfamily. BRSKs are crucial for establishing neuronal polarity, and BRSK1 has also been shown to regulate neurotransmitter release presynaptically. BRSK1 is a serine-threonine kinases specifically expressed in the brain, and activated by LKB1-mediated phosphorylation of a threonine residue at their T-loop. . It has 3 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: BRSK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR