Anti-TULP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12668-1-AP-150UL
10096-504EA 402 USD
10096-504
Anti-TULP2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, N/A

Type: Primary
Antigen: TULP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR