Anti-P4HA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12658-1-AP-150UL
10091-684EA 459.7 USD
10091-684
Anti-P4HA1 Rabbit Polyclonal Antibody
Antibodies
P4HA1(Prolyl 4-hydroxylase subunit alpha-1) is also named as P4HA and belongs to the P4HA family ,which play a central role in collagen synthesis. P4HA1 catalyzes the post-translational formation of 4-hydroxyproline in -Xaa-Pro-Gly- sequences in collagens and other proteins. The gene encodes a polypeptide of 517 amino acid residues and a signal peptide of 17 amino acids and the full length protein has two glycosylation sites(uniprot). It can exsit as a heteromer, dimer or tetramer(GENATLAS). P4HA1 has 3 isoforms produced by alternative splicing with the molecular weight of 61-63 kDa and 57-59 kDa.

Western Blot:A431 Cells, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200; IP:A431 Cells, 1:200-1:2000

Type: Primary
Antigen: P4HA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR