Anti-TCIRG1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12649-1-AP-150UL
10095-732EA 459.7 USD
10095-732
Anti-TCIRG1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HL-60 Cells, 1:200-1:2000; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: TCIRG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR