Anti-GNAO1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12635-1-AP-150UL
10087-682EA 453 USD
10087-682
Anti-GNAO1 Rabbit Polyclonal Antibody
Antibodies
IHC: Human gliomas Tissue, 1:20-1:200; Western Blot:rat Brain Tissue, 1:200-1:2000; IP: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: GNAO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR