Anti-LAMP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12632-1-AP-150UL
10089-496EA 402 USD
10089-496
Anti-LAMP3 Rabbit Polyclonal Antibody
Antibodies
LAMP3 (lysosome-associated membrane protein 3), also known as DC-LAMP (DC-lysosome-associated membrane glycoprotein), TSC403 or CD208, is a highly glycosylated lysosomal membrane protein that belongs to the LAMP family. The mature glycoprotein has an apparent molecular mass of 70-90 kDa, which is considerably larger than its predicted mass of 44 kDa . LAMP3 is expressed in lung type II pneumocytes, lymphoid organs and dendritic cells. It might change the lysosome function after the transfer of peptide-MHC class II molecules to the surface of dendritic cells. LAMP3 mRNA is up-regulated in some human cancers . LAMP3 overexpression is associated with an enhanced metastatic potential and may be a prognostic factor for cervical cancer .

Western Blot: Mouse lung Tissue, 1:500-1:5000; IHC: Human lung, 1:20-1:200

Type: Primary
Antigen: LAMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR