Anti-PLSCR4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12630-1-AP-150UL
10092-498EA 402 USD
10092-498
Anti-PLSCR4 Rabbit Polyclonal Antibody
Antibodies
PLSCR4 (Phospholipid scramblase 4) belongs to the phospholipid scramblase family which is thought to mediate the loss of lipid asymmetry. Elimination of this lipid bilayer asymmetry is an important physiological process and appears to occur as a result of elevated intracellular calcium concentration, or occur with cell activation, injury, and apoptosis.PLSCR4 also plays a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system.

Western Blot: Human Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: PLSCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR