Anti-Calmegin Rabbit Polyclonal Antibody

Supplier: Proteintech
12629-1-AP-150UL
10083-980EA 459.7 USD
10083-980
Anti-Calmegin Rabbit Polyclonal Antibody
Antibodies
Calmegin, belonging to the calreticulin family, is a testis-specific molecular chaperon of the endoplasmic reticulum playing a key role in spermatogenesis. Western blot analysis of mouse testis showed a single band with an apparent molecular mass of 93 kDa.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: Calmegin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR