Anti-PPP2R1B Rabbit Polyclonal Antibody

Supplier: Proteintech
12621-1-AP-150UL
10092-708EA 402 USD
10092-708
Anti-PPP2R1B Rabbit Polyclonal Antibody
Antibodies
Western Blot:A2780 Cells, 1:500-1:5000

Type: Primary
Antigen: PPP2R1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR