Anti-CDH13 Rabbit Polyclonal Antibody

Supplier: Proteintech
12618-1-AP-150UL
10083-948EA 402 USD
10083-948
Anti-CDH13 Rabbit Polyclonal Antibody
Antibodies
Cadherins are a family of transmembrane glycoproteins that mediate calcium-dependent cell-cell adhesion and play an important role in the maintenance of normal tissue architecture. Cadherin-13 (CDH13, also known as H-cadherin or T-cadherin) is an atypical member of cadherin family, lacking transmembrane and cytosolic domains and anchored to the exterior surface of the plasma membrane via a GPI anchor. CDH13 is thought to affect cellular behavior largely through its signaling properties. It is highly expressed in heart, and in the CNS, CDH13 is expressed in cerebral cortex, medulla, hippocampus, amygdala, thalamus and substantia nigra. It is often down-regulated in cancerous cells. The CDH13 cDNA encodes a 713-amino acid protein, which has a calculated molecular weight of 78 kDa. Studies have detected variable bands on Western blots, including 105 kDa (mature form), 130 kDa (precursor), and 45 kDa, which may be a post-translational processing variant of CDH13 .

Western Blot: Mouse Heart Tissue, 1:200-1:2000; IHC: Human endometrial cancer Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: CDH13
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR