Anti-TRPC4AP Rabbit Polyclonal Antibody

Supplier: Proteintech
12606-1-AP-150UL
10095-918EA 402 USD
10095-918
Anti-TRPC4AP Rabbit Polyclonal Antibody
Antibodies
TRPC4AP, also named as C20orf188 and TRRP4AP, is a substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex required for cell cycle control. TRPC4AP participates in the activation of NFKB1 in response to ligation of TNFRSF1A, possibly by linking TNFRSF1A to the IKK signalosome. The calculated molecular weight of TRPC4AP is 91 kDa. Western blot analysis detected a protein of about 82 kDa expressed in several mouse cell lines and in a human promonocytic cell line.

IHC: Human Prostate Cancer Tissue, 1:20-1:200; IP: Mouse Heart Tissue, 1:200-1:1000; Western Blot: Human Heart Tissue, 1:200-1:1000

Type: Primary
Antigen: TRPC4AP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR