Anti-CPB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12600-1-AP-150UL
10085-100EA 459.7 USD
10085-100
Anti-CPB1 Rabbit Polyclonal Antibody
Antibodies
PASP(pancreas-specific protein) is also named as CPB1, CPB, PCPB and belongs to the peptidase M14 family. This protein modulates the plasminogen system both in the matrix and on the cell surface, thus leading to the inhibition of endothelial cell movement and tube formation. It is initially described as a pancreatic metallocarboxypeptidase and is a marker for acute pancreatitis. The deduced 416-amino acid preproenzyme has a 15-amino acid signal peptide and a 95-amino acid activation peptide.

Western Blot: Mouse pancreas Tissue, 1:500-1:5000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR