Anti-CTBS Rabbit Polyclonal Antibody

Supplier: Proteintech
12599-1-AP-150UL
10085-298EA 402 USD
10085-298
Anti-CTBS Rabbit Polyclonal Antibody
Antibodies
CTBS(Di-N-acetylchitobiase (chitobiase)) also named as CTB, is a lysosomal exoglycosidase that splits the GlcNAcβ-D-(l-4)GlcNAc chitobiose core of asparagine-linked glycoproteins. Chitobiase has been purified from human liver and kidney and rat liver. Both the human and rat enzymes are approximately 40-45 kDa and require a free reducing end GlcNAc for activity. This full length protein has a signal peptide and four glycosylation sites.

Western Blot: MCF7 Cells, 1:200-1:1000; IHC: Human Prostate Cancer Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: CTBS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR