Anti-Cysteine dioxygenase type 1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12589-1-AP-150UL
10084-540EA 402 USD
10084-540
Anti-Cysteine dioxygenase type 1 Rabbit Polyclonal Antibody
Antibodies
CDO1(cysteine dioxygenase type 1) is also named as CDO and belongs to the cysteine dioxygenase family. It is an enzyme that adds molecular oxygen to the sulfur of cysteine, converting the thiol to a sulfinic acid known as cysteinesulfinic acid (3-sulfinoalanine) and CDO1 is one of the most highly regulated metabolic enzymes responding to diet. The expression of CDO can significantly decrease the level of intracellular cysteine and this reduction is also associated with a decrease in total glutathione levels.

Western Blot: Mouse Liver Tissue, 1:200-1:2000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: Cysteine dioxygenase type 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR