Anti-CRABP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12588-1-AP-150UL
10085-154EA 459.7 USD
10085-154
Anti-CRABP1 Rabbit Polyclonal Antibody
Antibodies
The cellular retinoic acid-binding proteins including CRABP1 and CRABP2 bind retinoic acid (RA), an important regulator of cell growth and differentiation, thus play an important role in RA-mediated differentiation and proliferation processes. It has been reported that CRABP1 influences the biological effects of RA in cell-selective manners by enhancing the physiological function of RA in keratinocytes or inhibiting RA-induced differentiation of neuroblastoma cells.

Western Blot: Human spleen Tissue, 1:300-1:1500

Type: Primary
Antigen: CRABP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR