Anti-PDCD4 Rabbit Polyclonal Antibody

Supplier: Proteintech
12587-1-AP-150UL
10091-976EA 402 USD
10091-976
Anti-PDCD4 Rabbit Polyclonal Antibody
Antibodies
Programmed cell death 4 (Pdcd4) is a novel tumor suppressor that inhibits translation, progression and invasion. It was first identified as being differnetially upregulated during apoptosis. Pdcd4 interferes with the activity of the eukaryotic initiation factor (eIF) 4A by displacing the scaffold protein eIF4G from its binding to the RNA helicase eIF4A.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human breast cancer Tissue, 1:20-1:200; FC: MCF-7 Cells, N/A

Type: Primary
Antigen: PDCD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR