Anti-THAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12584-1-AP-150UL
10095-436EA 402 USD
10095-436
Anti-THAP1 Rabbit Polyclonal Antibody
Antibodies
THAP1 belongs to the THAP1 family. It is a DNA-binding transcription regulator that regulates endothelial cell proliferation and G1/S cell-cycle progression. THAP1 may also have pro-apoptopic activity by potentiating both serum-withdrawal and TNF-induced apoptosis. Mutations in THAP1 have been associated with dystonia 6. THAP1 encodes a transcription factor with mostly unknown targets. It regulates the expression of DYT1 (TOR1A), another dystonia-causing gene. After characterization of the TOR1A promoter, THAP1 binds to the core promoter of TOR1A. Wild type THAP1 represses the expression of TOR1A, whereas dystonia 6-associated mutant THAP1 results in decreased repression of TOR1A. Catalog#12584-1-AP is a rabbit polyclonal antibody raised against full-length of huamn THAP1.

Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human Colon cancer Tissue, 1:20-1:200

Type: Primary
Antigen: THAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR