Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EDIL3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12580-1-AP
EGF-like repeats and discoidin I-like domain-containing protein 3 (EDIL3), also named as DEL1 or integrin-biding protein DEL1, is a 52-kDa extracellular matrix protein produced by endothelial cells in embryos . It is composed of three EGF repeats and two discoidin I-like domains. The second EGF repeat contains an RGD motif. EDIL3 promotes adhesion of endothelial cells through interaction with the alpha-v/beta-3 integrin receptor. It may be involved in vascular remodeling during angiogenesis . EDIL3 has also been reported to be an endogenous inhibitor of inflammatory cell recruitment by interfering with the integrin LFA-1-dependent leukocyte-endothelial adhesion .

Western Blot: Human lung Tissue, 1:1000-1:10000; IHC: Human pancreas cancer Tissue, 1:20-1:200

Type: Primary
Antigen: EDIL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
150 µL 12580-1-AP 10086-264 Each (150µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol EDIL3
Antigen Name EGF-Like Repeats And Discoidin I-Like Domains 3
Reactivity Human, Mouse, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody Type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
Antigen Synonyms Developmentally-Regulated Endothelial Cell Locus 1 Protein, Integrin-Binding Protein DEL1, Developmental Endothelial Locus-1, DEL1
Immunogen Recombinant Protein
Purification Antigen affinity Purification
Storage Buffer PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
Shipping Temperature Store at -20°C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping