Anti-TSP50 Rabbit Polyclonal Antibody

Supplier: Proteintech
12574-1-AP-150UL
10096-416EA 459.7 USD
10096-416
Anti-TSP50 Rabbit Polyclonal Antibody
Antibodies
TSP50 (Testis-specific protease-like protein 50) is also named as PRSS50, CT20 and belongs to the peptidase S1 family. It can enhance the expression of NFKB target genes involved in cell proliferation, and promotes cell proliferation, at least partially, through activation of the NFKB signalling pathway. There are two protein bands of 37 kDa and 43 kDa to be detected in human testes that the the difference is most likely caused by posttranslational proteolytic cleavage. The full length protein has a signal peptide with 39 amino acids and two glycosylation sites.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: MCF-7 Cells, 1:10-1:100

Type: Primary
Antigen: TSP50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR