Anti-EPAC1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12572-1-AP-150UL
10082-252EA 402 USD
10082-252
Anti-EPAC1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:CHO Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200; IF:A431 Cells, 1:10-1:100

Type: Primary
Antigen: EPAC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR