Anti-PBX3 Rabbit Polyclonal Antibody

Supplier: Proteintech
12571-1-AP-150UL
10091-892EA 402 USD
10091-892
Anti-PBX3 Rabbit Polyclonal Antibody
Antibodies
PBX3, also named as Pre-B-cell leukemia transcription factor 3, is a 434 amino acid protein ,which contains 1 homeobox DNA-binding domain and belongs to the TALE/PBX homeobox family. PBX3 localizes in the nucleus and as a transcriptional activator that binds the sequence 5'-ATCAATCAA-3'. PBX3 is ubiquitously expressed in many tissues.

Western Blot: Human lung Tissue, 1:200-1:2000; IHC: Human lung cancer Tissue, 1:20-1:200

Type: Primary
Antigen: PBX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR