Anti-SCFD1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12569-1-AP-150UL
10094-176EA 459.7 USD
10094-176
Anti-SCFD1 Rabbit Polyclonal Antibody
Antibodies
SCFD1, also named as C14orf163, KIAA0917, STXBP1L2, Sly1p and SLY1, belongs to the STXBP/unc-18/SEC1 family. It is a component of the syntaxin-5 SNARE complex which regulates the early secretory pathway of eukaryotic cells at the level of endoplasmic reticulum (ER) to Golgi transport. SCFD1 plays a role in SNARE-pin assembly and Golgi-to-ER retrograde transport via its interaction with COG4. It is involved in vesicular transport between the endoplasmic reticulum and the Golgi.

Western Blot:A549 Cells, 1:500-1:5000; IHC: Human endometrial cancer Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: SCFD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR