Anti-AMPK gamma2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12568-1-AP-150UL
10082-862EA 402 USD
10082-862
Anti-AMPK gamma2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Heart Tissue, 1:200-1:2000; IHC: Human Placenta Tissue, 1:20-1:200; IF: HeLa Cells, 1:20-1:200

Type: Primary
Antigen: AMPK gamma2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR