![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SELK |
Antigen Name | Selenoprotein K |
Reactivity | Rat, Mouse, Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
Antigen Synonyms | SelK, HSPC297, HSPC030, SELK, Selenoprotein K |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage Buffer | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
Shipping Temperature | Store at -20°C |