Fucosyltransferase 8 (FUT8) is GDP-fucose-glycoprotein fucosyltransferase and is localized in Golgi apparatus. FUT8 is involved in protein glycosylation by catalyzing the addition of fucose in alpha 1-6 linkage to the first GlcNAc residue, next to the peptide chains in N-glycans. FUT8 may be involved in human aging by regulating protein N-glycosylation and in the molecular pathogenesis of human pulmonary emphysema (PE).
Western Blot: HEK-293 Cells, 1:500-1:5000; IHC: Human lung cancer Tissue, 1:20-1:200
Type: Primary
Antigen: FUT8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat