Anti-CPVL Rabbit Polyclonal Antibody

Supplier: Proteintech
12548-1-AP-150UL
10085-152EA 599.76 USD
10085-152
Anti-CPVL Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:1000-1:10000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: CPVL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR