Anti-SQLE Rabbit Polyclonal Antibody

Supplier: Proteintech
12544-1-AP-150UL
10095-140EA 537.7 USD
10095-140
Anti-SQLE Rabbit Polyclonal Antibody
Antibodies
SQLE, also named as ERG1, SE and SM, belongs to the squalene monooxygenase family. It catalyzes the first oxygenation step in cholesterol synthesis, acting on squalene before cyclization into the basic steroid structure. SQLE may serve as a flux-controlling enzyme beyond 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR, considered as rate limiting). It is also posttranslationally regulated by cholesterol-dependent proteasomal degradation. The MW of SQLE is about 64kd.

Western Blot:A549 Cells, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:20-1:200; IP:A549 Cells, 1:200-1:1000

Type: Primary
Antigen: SQLE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR