Anti-RND3 Rabbit Polyclonal Antibody

12534-1-AP
10093-780EA 349.28 USD
10093-780
Anti-RND3 Rabbit Polyclonal Antibody
Western Blot: Mouse Skeletal Muscle Tissue, 1:200-1:2000; IHC: Human Liver cancer, 1:20-1:200

Type: Primary
Antigen: RND3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR