Anti-GAGE2D Rabbit Polyclonal Antibody

Supplier: Proteintech
12532-1-AP-150UL
10087-316EA 402 USD
10087-316
Anti-GAGE2D Rabbit Polyclonal Antibody
Antibodies
GAGE2D, also named as GAGE8 and CT4.8, is expressed in testis and tumor tissues.

Western Blot:K-562 Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: GAGE2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR