|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PLAA |
Antigen Name | Phospholipase A2-Activating Protein |
Reactivity | Human, Mouse, Rat |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
Immunogen | Recombinant Protein |
Purification | Antigen affinity Purification |
Storage Buffer | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
Shipping Temperature | Store at -20°C |