Anti-SULT1E1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12522-1-AP-150UL
10095-444EA 402 USD
10095-444
Anti-SULT1E1 Rabbit Polyclonal Antibody
Antibodies
SULT1E1, also known as estrogen sulfotransferase, catalyzes the sulfation of endogenous estrogens as well as xenobiotic estrogen-like chemicals, converting them into the inactive form. SULT1E1 is a 33-35 kDa cytosolic protein that has been detected in human hepatic and nonhepatic tissues (17035602). SULT1E1 is expressed in breast and endometrial tissues, and some studies have investigated its implication in tumor development. Higher expression of SULT1E1 was observed in breast cancer tissues compared with normal breast tissues (22380844). This antibody detected a major band around 35 kD in lysates of mouse kidney, human liver and so on.

Western Blot: Human lung Tissue, 1:200-1:2000

Type: Primary
Antigen: SULT1E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR