Anti-TRUB1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12520-1-AP-150UL
10095-718EA 459.7 USD
10095-718
Anti-TRUB1 Rabbit Polyclonal Antibody
Antibodies
TRUB1 gene encodes a 349-amino acid product, with a VFAVHKPKGPTSA box in positions 71-83 corresponding to motif I of the TruB family (probably involved in conserving protein structure). This protein may be responsible for synthesis of pseudouridine from uracil in transfer RNAs.

Western Blot: Mouse Liver Tissue, 1:200-1:1000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: TRUB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR