Anti-NUFIP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
12515-1-AP-150UL
10091-390EA 459.7 USD
10091-390
Anti-NUFIP1 Rabbit Polyclonal Antibody
Antibodies
Fragile X syndrome, the most common cause of inherited mental retardation, is caused by the absence of FMRP (Fragile X Mental Retardation Protein). FMRP is an RNA binding protein reported to be involved in translational control, notably at postsynaptic sites of protein synthesis as a part of a multiprotein/mRNA complex. NUFIP1 is one of the several FMRP-interacting proteins. NUFIP can act as a pol II-specific basal transcriptional activator in vitro and when ectopically overexpressed in vivo. NUFIP can directly activate promoters by enhancing the ATP-dependent release of hyperphosphorylated form of pol II from open transcription complexes.

Western Blot:A375 Cells, 1:500-1:5000; IHC: Human ovary tumor, 1:20-1:200

Type: Primary
Antigen: NUFIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR