Anti-RGS3 Rabbit Polyclonal Antibody

12501-1-AP
10093-626EA 356.38 USD
10093-626
Anti-RGS3 Rabbit Polyclonal Antibody
IHC: Human gliomas Tissue, 1:20-1:200; Western Blot:Transfected HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: RGS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR