Anti-ELF2 Rabbit Polyclonal Antibody

Supplier: Proteintech
12499-1-AP-150UL
10086-468EA 459.7 USD
10086-468
Anti-ELF2 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A2780 Cells, 1:500-1:5000

Type: Primary
Antigen: ELF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR